Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012547.1 | NP_036679.1 | 6 | 1098 | ||
XM_008766190.1 | XP_008764412.1 | 5 | 1115 | |||
GenBank mRNA | M32241.1 | - | 1 | 29 | ||
X53278.1 | - | 6 | 878 | |||
X56065.1 | - | 6 | 1098 |
Set Membership: |
G-protein coupled receptor |