Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030826.3 | NP_110453.3 | 2 | 625 | ||
GenBank mRNA | BC058438.1 | - | 2 | 597 | 6918849 | |
BI281978.1 | - | 1 | 270 | |||
FQ211052.1 | - | 2 | 622 | |||
FQ227523.1 | - | 2 | 654 | |||
M21210.1 | - | 2 | 631 |