Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012576.2 | NP_036708.2 | 5 | 1939 | ||
XM_008772065.1 | XP_008770287.1 | 5 | 1940 | |||
XM_008772066.1 | XP_008770288.1 | 6 | 1990 | |||
GenBank mRNA | AB115420.1 | - | 5 | 1926 | ||
AF455050.1 | - | 5 | 1930 | |||
AY066016.2 | - | 5 | 1934 | |||
CB699574.1 | - | 1 | 141 | |||
M14053.1 | - | 5 | 1997 | |||
Y12264.1 | - | 5 | 1951 |