Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031509.2 | NP_113697.1 | 7 | 889 | ||
GenBank mRNA | AF111160.1 | - | 7 | 889 | ||
BC059128.1 | - | 7 | 705 | 6922234 | ||
BC088127.1 | - | 7 | 689 | 7322480 | ||
FJ179398.1 | - | 6 | 650 | |||
FQ209644.1 | - | 7 | 705 | |||
FQ211002.1 | - | 7 | 700 | |||
FQ211022.1 | - | 7 | 701 | |||
FQ213952.1 | - | 7 | 840 | |||
FQ218347.1 | - | 7 | 701 | |||
FQ218448.1 | - | 7 | 698 | |||
FQ218456.1 | - | 7 | 701 | |||
FQ218470.1 | - | 7 | 703 | |||
FQ218508.1 | - | 7 | 700 | |||
FQ219165.1 | - | 7 | 701 | |||
FQ219196.1 | - | 7 | 701 | |||
FQ219451.1 | - | 7 | 709 | |||
FQ219713.1 | - | 7 | 701 | |||
FQ226984.1 | - | 7 | 703 | |||
FQ229239.1 | - | 7 | 701 | |||
K01932.1 | - | 7 | 694 | |||
S72505.1 | - | 7 | 799 | |||
X78848.1 | - | 7 | 799 |
Gene Symbol: LOC102550391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244659.2 | XP_006244721.2 | 884 |