Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012599.2 | NP_036731.2 | 5 | 729 | ||
GenBank mRNA | BC088159.1 | - | 5 | 729 | 7326244 | |
FQ209936.1 | - | 5 | 745 | |||
FQ211562.1 | - | 1 | 283 | |||
FQ218582.1 | - | 5 | 757 | |||
FQ219753.1 | - | 1 | 283 |