Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139253.1 | NP_640346.1 | 4 | 426 | ||
XM_006239710.2 | XP_006239772.1 | 4 | 465 | |||
XM_006239711.2 | XP_006239773.1 | 4 | 486 | |||
GenBank mRNA | AF475282.1 | - | 4 | 426 | ||
BC071179.1 | - | 4 | 420 | 7100216 |