Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144738.2 | NP_653339.2 | 15, 16 | 1756 | ||
GenBank mRNA | BC091230.1 | - | 15, 16 | 1756 | 7321805 |
Gene Symbol: LOC100911867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749429.3 | XP_003749477.2 | 15, 16 | 1820 |