Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169120.1 | NP_001162591.1 | 3 | 457 | ||
XM_006231127.2 | XP_006231189.1 | 3 | 711 |