Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012625.1 | NP_036757.1 | 2 | 323 | ||
GenBank mRNA | FQ214578.1 | - | 2 | 320 | ||
M29712.1 | - | 2 | 307 |
Gene Symbol: Parpbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166676.1 | NP_001160148.1 | 12 | 3281 | ||
XM_006241188.2 | XP_006241250.1 | 11 | 3108 | |||
XM_006241189.2 | XP_006241251.1 | 11 | 3946 | |||
XM_006241190.2 | XP_006241252.1 | 11 | 3192 | |||
XM_008765209.1 | XP_008763431.1 | 11 | 3457 | |||
XM_008765210.1 | XP_008763432.1 | 11 | 3454 | |||
XM_008765211.1 | XP_008763433.1 | 11 | 3104 | |||
GenBank mRNA | AF303035.1 | - | 12 | 3281 |