Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CA339848.1 | - | 2 | 288 | ||
J00791.1 | - | 2 | 270 | |||
M25590.1 | - | 2 | 270 | |||
X01114.1 | - | 2 | 321 |
Gene Symbol: LOC103691965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762550.1 | XP_008760772.1 | 2 | 267 |
Gene Symbol: LOC103694889 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773870.1 | XP_008772092.1 | 312 |