Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270664.1 | NP_001257593.1 | 13, 14 | 766 | ||
NM_001270665.1 | NP_001257594.1 | 15, 16 | 799 | |||
NM_001270671.1 | NP_001257600.1 | 14, 15 | 781 | |||
NM_001270672.1 | NP_001257601.1 | 13, 14 | 772 | |||
NM_001270673.1 | NP_001257602.1 | 14, 15 | 784 | |||
NM_001270677.1 | NP_001257606.1 | 12, 13 | 754 | |||
NM_031532.2 | NP_113720.1 | 14, 15 | 787 | |||
XM_006230681.2 | XP_006230743.1 | 13, 14 | 729 | |||
XM_006230683.2 | XP_006230745.1 | 12, 13 | 717 | |||
XM_006230685.2 | XP_006230747.1 | 11, 12 | 699 | |||
XM_008760080.1 | XP_008758302.1 | 16, 17 | 833 | |||
XM_008760082.1 | XP_008758304.1 | 15, 16 | 783 | |||
XM_008760083.1 | XP_008758305.1 | 15, 16 | 780 | |||
XM_008760084.1 | XP_008758306.1 | 14, 15 | 806 | |||
XM_008760085.1 | XP_008758307.1 | 14, 15 | 771 | |||
XM_008760086.1 | XP_008758308.1 | 13, 14 | 753 | |||
GenBank mRNA | DQ273678.1 | - | 14, 15 | 683 | ||
FQ214650.1 | - | 14, 15 | 747 | |||
FQ214722.1 | - | 13, 14 | 734 | |||
FQ214833.1 | - | 14, 15 | 749 | |||
FQ215106.1 | - | 13, 14 | 728 | |||
FQ215114.1 | - | 13, 14 | 734 | |||
FQ215252.1 | - | 14, 15 | 746 | |||
FQ215316.1 | - | 13, 14 | 748 | |||
FQ215327.1 | - | 15, 16 | 761 | |||
FQ215598.1 | - | 15, 16 | 757 | |||
FQ215602.1 | - | 13, 14 | 761 | |||
FQ215694.1 | - | 13, 14 | 766 | |||
FQ215723.1 | - | 15, 16 | 800 | |||
FQ215881.1 | - | 15, 16 | 761 | |||
FQ215885.1 | - | 15, 16 | 749 | |||
FQ215897.1 | - | 14, 15 | 774 | |||
FQ215955.1 | - | 13, 14 | 728 | |||
FQ216059.1 | - | 15, 16 | 756 | |||
FQ216131.1 | - | 13, 14 | 759 | |||
FQ216208.1 | - | 14, 15 | 746 | |||
FQ216324.1 | - | 13, 14 | 728 | |||
FQ216367.1 | - | 13, 14 | 728 | |||
FQ216417.1 | - | 14, 15 | 748 | |||
FQ216467.1 | - | 13, 14 | 728 | |||
FQ216561.1 | - | 10, 11 | 728 | |||
FQ216711.1 | - | 13, 14 | 765 | |||
FQ216750.1 | - | 10, 11 | 619 | |||
FQ216763.1 | - | 11, 12 | 719 | |||
FQ216784.1 | - | 13, 14 | 734 | |||
FQ216866.1 | - | 15, 16 | 761 | |||
FQ217147.1 | - | 13, 14 | 727 | |||
FQ217651.1 | - | 11 | ||||
FQ217900.1 | - | 14, 15 | 774 | |||
FQ223840.1 | - | 13, 14 | 728 | |||
FQ224794.1 | - | 13, 14 | 728 | |||
FQ224813.1 | - | 15, 16 | 761 | |||
FQ224892.1 | - | 14, 15 | 749 |