Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012678.2 | NP_036810.1 | 8 | 753 | ||
GenBank mRNA | FQ225054.1 | - | 764 | |||
FQ225082.1 | - | 764 | ||||
J02780.1 | - | 8 | 756 |