Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012695.3 | NP_036827.3 | 2, 3 | 385 | ||
GenBank mRNA | FQ209484.1 | - | 2, 3 | 260 | ||
FQ209563.1 | - | 2, 3 | 260 | |||
FQ218388.1 | - | 2, 3 | 260 | |||
FQ218712.1 | - | 2, 3 | 260 | |||
FQ219267.1 | - | 2, 3 | 260 | |||
FQ219345.1 | - | 2, 3 | 260 | |||
M33329.1 | - | 2, 3 | 385 | |||
X63410.1 | - | 2, 3 | 385 |
Gene Symbol: LOC100360574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759026.1 | XP_008757248.1 | 2, 3 | 348 |
Gene Symbol: Sult2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131903.1 | NP_571978.1 | 2, 3 | 385 | ||
GenBank mRNA | D14987.1 | - | 2, 3 | 385 | ||
D14988.1 | - | 2, 3 | 385 | |||
FQ209430.1 | - | 2, 3 | 260 | |||
FQ209460.1 | - | 2, 3 | 260 | |||
FQ210003.1 | - | 2, 3 | 260 | |||
FQ218232.1 | - | 2, 3 | 260 | |||
FQ218962.1 | - | 2, 3 | 260 | |||
FQ219256.1 | - | 2, 3 | 260 | |||
FQ219272.1 | - | 2, 3 | 281 |