Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053018.1 | NP_444177.1 | 4 | 350 | ||
GenBank mRNA | BC161804.1 | - | 4 | 325 | 9101765 | |
CF111078.1 | - | 4 | 359 | |||
FQ212497.1 | - | 4 | 374 | |||
FQ213039.1 | - | 4 | 399 | |||
FQ213302.1 | - | 4 | 368 | |||
FQ215636.1 | - | 4 | 362 | |||
FQ216402.1 | - | 4 | 399 | |||
FQ216770.1 | - | 4 | 386 | |||
FQ219799.1 | - | 4 | 373 | |||
FQ220476.1 | - | 4 | 374 | |||
FQ224453.1 | - | 4 | 374 | |||
FQ229965.1 | - | 4 | 421 | |||
KC162232.1 | - | 289 | ||||
KC162233.1 | - | 3 | 336 | |||
X76489.1 | - | 4 | 289 |