Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012721.2 | NP_036853.2 | 3, 4 | 493 | ||
XM_008768851.1 | XP_008767073.1 | 3, 4 | 683 | |||
XM_008768852.1 | XP_008767074.1 | 3, 4 | 368 | |||
GenBank mRNA | X92070.1 | - | 3, 4 | 374 | ||
X97376.1 | - | 3, 4 | 504 |