Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012736.1 | NP_036868.1 | 12 | 1657 | ||
GenBank mRNA | BC083565.1 | - | 12 | 1615 | 7134834 | |
FQ223746.1 | - | 1913 | ||||
U08027.1 | - | 12 | 1636 | |||
X78593.1 | - | 12 | 1657 |