Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102418.1 | NP_001095888.1 | 7 | 988 | ||
NM_012741.1 | NP_036873.1 | 7 | 889 | |||
GenBank mRNA | BC088155.1 | - | 7 | 988 | 7369450 | |
L29428.1 | - | 7 | 889 | |||
M11884.1 | - | 5 | 543 |