Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031546.1 | NP_113734.1 | 6 | 848 | ||
GenBank mRNA | AB037934.1 | - | 6 | 848 | ||
BC078794.1 | - | 6 | 843 | 7126233 | ||
D38467.1 | - | 6 | 842 | |||
FQ219778.1 | - | 6 | 851 | |||
X69021.1 | - | 6 | 837 |