Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031551.1 | NP_113739.1 | 11, 12 | 1076 | ||
XM_006229551.1 | XP_006229613.1 | 9, 10 | 1498 | |||
XM_006229552.2 | XP_006229614.1 | 10, 11 | 1081 | |||
XM_006229553.2 | XP_006229615.1 | 11, 12 | 1101 | |||
XM_006229554.1 | XP_006229616.1 | 10, 11 | 1028 | |||
GenBank mRNA | BC083688.1 | - | 11, 12 | 1076 | 7189278 | |
FQ213717.1 | - | 11, 12 | 955 | |||
FQ214482.1 | - | 11, 12 | 1043 | |||
FQ214526.1 | - | 11, 12 | 1042 | |||
FQ214664.1 | - | 11, 12 | 1017 | |||
FQ214718.1 | - | 11, 12 | 1093 | |||
FQ214896.1 | - | 11, 12 | 1026 | |||
FQ216691.1 | - | 11, 12 | 1032 | |||
FQ217943.1 | - | 1, 2 | 103 | |||
FQ226989.1 | - | 10, 11 | 953 | |||
X74125.1 | - | 11, 12 | 1002 |