Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109996.1 | NP_001103466.1 | 3 | 213 | ||
NM_012824.2 | NP_036956.1 | 2 | 248 | |||
GenBank mRNA | AY327505.1 | - | 3 | 230 | ||
BC098670.1 | - | 2 | 187 | 7372566 | ||
FQ210440.1 | - | 3 | 213 | |||
FQ210757.1 | - | 3 | 202 | |||
FQ211211.1 | - | 3 | 214 | |||
FQ212377.1 | - | 3 | 213 | |||
FQ223744.1 | - | 3 | 213 | |||
X15512.1 | - | 2 | 248 |
Gene Symbol: LOC100911905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752245.2 | XP_003752293.2 | 3 | 247 |