Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012840.3 | NP_036972.1 | 505 | |||
GenBank mRNA | M20623.1 | - | 2 | 335 |