Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012876.1 | NP_037008.1 | 1 | 83 | ||
GenBank mRNA | BC058150.1 | - | 1 | 81 | 6922365 | |
FQ209501.1 | - | 1 | 77 | |||
FQ210507.1 | - | 1 | 77 | |||
FQ211411.1 | - | 1 | 92 | |||
FQ211740.1 | - | 1 | 77 | |||
FQ216880.1 | - | 1 | 77 | |||
FQ217140.1 | - | 1 | 77 | |||
FQ217213.1 | - | 1 | 77 | |||
FQ217476.1 | - | 1 | 77 | |||
FQ218006.1 | - | 1 | 77 | |||
FQ218037.1 | - | 1 | 78 | |||
FQ218769.1 | - | 1 | 77 | |||
FQ221030.1 | - | 1 | 77 | |||
FQ221251.1 | - | 1 | 77 | |||
FQ221298.1 | - | 1 | 77 | |||
FQ221307.1 | - | 1 | 74 | |||
FQ221360.1 | - | 1 | 77 | |||
FQ221363.1 | - | 1 | 77 | |||
FQ221457.1 | - | 1 | 77 | |||
FQ221523.1 | - | 1 | 77 | |||
FQ221587.1 | - | 1 | 78 | |||
FQ221640.1 | - | 1 | 77 | |||
FQ221707.1 | - | 1 | 77 | |||
FQ221875.1 | - | 1 | 77 | |||
FQ221942.1 | - | 1 | 77 | |||
FQ222143.1 | - | 1 | 77 | |||
FQ222293.1 | - | 1 | 77 | |||
FQ222368.1 | - | 1 | 77 | |||
FQ222566.1 | - | 1 | 77 | |||
FQ222711.1 | - | 1 | 77 | |||
FQ222937.1 | - | 1 | 77 | |||
FQ223071.1 | - | 1 | 77 | |||
FQ223222.1 | - | 1 | 77 | |||
FQ223289.1 | - | 1 | 77 | |||
FQ223378.1 | - | 1 | 77 | |||
FQ223402.1 | - | 1 | 77 | |||
FQ223735.1 | - | 1 | 114 | |||
FQ223885.1 | - | 1 | 77 | |||
FQ223975.1 | - | 1 | 77 | |||
FQ224325.1 | - | 1 | 77 | |||
FQ224400.1 | - | 1 | 77 | |||
FQ224465.1 | - | 1 | 77 | |||
FQ224476.1 | - | 1 | 77 | |||
FQ224629.1 | - | 1 | 84 | |||
FQ224661.1 | - | 1 | 78 | |||
FQ224820.1 | - | 1 | 77 | |||
FQ228434.1 | - | 1 | 77 | |||
FQ228730.1 | - | 1 | 77 | |||
X59051.1 | - | 1 | 83 |