Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033715.1 | NP_001028887.1 | 9, 10 | 765 | ||
NM_001033716.1 | NP_001028888.1 | 8, 9 | 696 | |||
NM_053295.2 | NP_445747.2 | 10, 11 | 879 | |||
XM_006231698.2 | XP_006231760.1 | 10, 11 | 1287 | |||
XM_006231699.2 | XP_006231761.1 | 9, 10 | 1012 | |||
GenBank mRNA | AF346597.1 | - | 6, 7 | 377 | ||
AF346598.1 | - | 8, 9 | 560 | |||
BC091239.1 | - | 9, 10 | 765 | 7324026 | ||
DQ186624.1 | - | 10, 11 | 1073 | |||
DQ186625.1 | - | 8, 9 | 890 | |||
DQ186626.1 | - | 9, 10 | 1004 | |||
DQ186627.1 | - | 9, 10 | 959 | |||
DQ186628.1 | - | 8, 9 | 920 | |||
DQ186629.1 | - | 7, 8 | 851 | |||
DQ287975.1 | - | 3, 4 | 751 | |||
Y13587.1 | - | 7, 8 | 458 | |||
Y13588.1 | - | 8, 9 | 572 | |||
Y13589.1 | - | 6, 7 | 389 | |||
Y13591.1 | - | 7, 8 | 458 |