Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpysl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105717.2 | NP_001099187.1 | 1, 2 | 266 | ||
GenBank mRNA | CB749736.1 | - | 1, 2 | 323 | ||
Z46882.1 | - | 1, 2 | 219 |