Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012990.1 | NP_037122.1 | 1 | 926 | ||
GenBank mRNA | U31203.1 | - | 1 | 413 |