Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013025.2 | NP_037157.2 | 3 | 306 | ||
GenBank mRNA | EF121994.1 | - | 3 | 237 | ||
EF121995.1 | - | 3 | 237 | |||
U06435.1 | - | 3 | 237 | |||
U22414.1 | - | 3 | 306 |