Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sepw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013027.3 | NP_037159.4 | 2, 3 | 144 | ||
GenBank mRNA | BC087625.1 | - | 2, 3 | 120 | 7316562 | |
BI279050.1 | - | 568 | ||||
DN931873.1 | - | 2, 3 | 166 | |||
FQ212381.1 | - | 2, 3 | 130 | |||
FQ212407.1 | - | 2, 3 | 124 | |||
FQ215130.1 | - | 2, 3 | 123 | |||
FQ215391.1 | - | 2, 3 | 125 | |||
FQ216964.1 | - | 2, 3 | 120 | |||
FQ217212.1 | - | 2, 3 | 125 | |||
FQ217426.1 | - | 2, 3 | 126 | |||
FQ217557.1 | - | 2, 3 | 126 | |||
FQ217567.1 | - | 2, 3 | 126 | |||
FQ217929.1 | - | 2, 3 | 126 | |||
FQ221681.1 | - | 2, 3 | 126 | |||
FQ223747.1 | - | 2, 3 | 126 | |||
FQ224556.1 | - | 2, 3 | 130 | |||
FQ224570.1 | - | 2, 3 | 126 | |||
FQ224606.1 | - | 2, 3 | 123 | |||
FQ224775.1 | - | 2, 3 | 126 | |||
U25264.1 | - | 2, 3 | 111 |