Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038597.2 | NP_001033686.1 | 1 | 461 | ||
NM_001038599.2 | NP_001033688.2 | 1 | 461 | |||
NM_001038600.2 | NP_001033689.1 | 1 | 461 | |||
NM_001038601.2 | NP_001033690.1 | 1 | 461 | |||
NM_013071.2 | NP_037203.1 | 1 | 461 | |||
XM_008758731.1 | XP_008756953.1 | 1 | 607 | |||
GenBank mRNA | AY225402.1 | - | 1 | 292 | ||
AY225403.1 | - | 1 | 292 | |||
AY309000.1 | - | 1 | 292 | |||
AY309002.1 | - | 1 | 286 | |||
AY309003.1 | - | 1 | 292 | |||
AY309004.1 | - | 1 | 292 | |||
D16349.1 | - | 1 | 437 | |||
EU024651.1 | - | 473 | ||||
EU024652.1 | - | 662 | ||||
EU340244.1 | - | 481 | ||||
EU340245.1 | - | 737 | ||||
EU340246.1 | - | 923 | ||||
FJ041289.1 | - | 1 | 292 | |||
HQ699463.1 | - | 1 | 292 | |||
L13069.1 | - | 1 | 463 | |||
L20684.1 | - | 1 | 284 | |||
L22455.1 | - | 1 | 423 | |||
U02083.1 | - | 1 | 464 | |||
U35424.1 | - | 1 | 323 |
Set Membership: |
G-protein coupled receptor |