Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col10a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001053056.6 | XP_001053056.5 | 3 | 5285 | ||
XM_008773017.1 | XP_008771239.1 | 2 | 5802 | |||
XM_008773018.1 | XP_008771240.1 | 3 | 5758 | |||
GenBank mRNA | AJ131848.1 | - | 1 | 677 |