Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013162.1 | NP_037294.1 | 3 | 281 | ||
GenBank mRNA | BC167099.1 | - | 3 | 282 | 7369314 | |
CO561550.1 | - | 3 | 281 | |||
DV725071.1 | - | 1 | 80 | |||
FQ218638.1 | - | 3 | 297 | |||
M10934.1 | - | 2 | 144 |