Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762559.1 | XP_008760781.1 | 21 | 4121 | ||
XM_008762560.1 | XP_008760782.1 | 22 | 4795 | |||
XM_008762561.1 | XP_008760783.1 | 21 | 4118 | |||
XM_008762562.1 | XP_008760784.1 | 21 | 4121 | |||
XM_008762563.1 | XP_008760785.1 | 21 | 4121 | |||
XM_008762564.1 | XP_008760786.1 | 21 | 4118 | |||
XM_008762565.1 | XP_008760787.1 | 21 | 4121 | |||
GenBank mRNA | AF309071.1 | - | 7 | 1143 | ||
AY079168.1 | - | 13 | 1893 |