Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134643.2 | NP_001128115.2 | 4 | 869 | ||
GenBank mRNA | AF525937.1 | - | 1 | 77 | ||
FQ084615.1 | - | 4 | 510 |