Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100487.1 | NP_001093957.1 | 2 | 182 | ||
GenBank mRNA | AF536558.3 | - | 1 | 48 | ||
AY834200.1 | - | 1 | 91 |