Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153474.1 | NP_703204.1 | 4 | 973 | ||
GenBank mRNA | AF323956.1 | - | 4 | 973 |
Set Membership: |
G-protein coupled receptor |