Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004442.1 | NP_001004442.1 | 3, 4 | 249 | ||
GenBank mRNA | BC082063.1 | - | 3, 4 | 249 | 7132589 | |
X79328.1 | - | 2, 3 | 189 |