Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173295.1 | NP_775417.1 | 3, 4 | 1039 | ||
GenBank mRNA | M13506.1 | - | 3, 4 | 1039 |
Gene Symbol: RGD1565664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770100.1 | XP_008768322.1 | 2, 3 | 267 |