Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246538.2 | XP_006246600.1 | 4 | 1218 | ||
XM_220559.8 | XP_220559.5 | 5 | 1044 |