Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnasek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137561.2 | NP_001131033.2 | 3 | 350 | ||
GenBank mRNA | BC169131.1 | - | 3 | 290 | 6922284 | |
CB719759.1 | - | 3 | 350 |