Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105803.2 | NP_001099273.2 | 3 | 687 | ||
GenBank mRNA | BI274683.1 | - | 1 | 589 | ||
CB326294.1 | - | 1 | 567 |