Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105815.1 | NP_001099285.1 | 5, 6 | 737 | ||
XM_006246935.2 | XP_006246997.1 | 4, 5 | 551 |