Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105824.1 | NP_001099294.1 | 14 | 1297 | ||
GenBank mRNA | BC166769.1 | - | 16 | 1873 | 7933997 |