Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105824.1 | NP_001099294.1 | 4 | 241 | ||
GenBank mRNA | BC166769.1 | - | 4 | 193 | 7933997 |
Gene Symbol: LOC103694865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773813.1 | XP_008772035.1 | 248 | |||
XM_008773814.1 | XP_008772036.1 | 420 |