Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Higd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105844.1 | NP_001099314.1 | 2 | 537 | ||
XM_008768263.1 | XP_008766485.1 | 3 | 375 | |||
XM_008768264.1 | XP_008766486.1 | 3 | 260 | |||
XM_008768265.1 | XP_008766487.1 | 3 | 403 |