Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105846.1 | NP_001099316.1 | 16, 17 | 2411 | ||
XM_006247498.2 | XP_006247560.1 | 19, 20 | 2811 | |||
XM_006247499.2 | XP_006247561.1 | 19, 20 | 2812 | |||
XM_006247500.2 | XP_006247562.1 | 18, 19 | 2793 | |||
XM_006247501.2 | XP_006247563.1 | 18, 19 | 2793 | |||
XM_006247502.2 | XP_006247564.1 | 19, 20 | 2781 | |||
XM_008768266.1 | XP_008766488.1 | 17, 18 | 2666 | |||
XM_008768267.1 | XP_008766489.1 | 17, 18 | 2637 |