Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smndc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025400.3 | NP_001020571.1 | 4 | 347 | ||
NM_001035234.2 | NP_001030311.1 | 4 | 509 | |||
XM_006231608.2 | XP_006231670.1 | 4 | 786 | |||
GenBank mRNA | BC097986.1 | - | 4 | 347 | 7444662 | |
BC098697.1 | - | 4 | 488 | 7386640 | ||
EV778425.1 | - | 4 | 509 | |||
FQ220088.1 | - | 3 | 290 |
Gene Symbol: LOC100363406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768448.1 | XP_008766670.1 | 2 | 472 |