Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2v2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183052.2 | NP_898875.1 | 2 | 182 | ||
XM_006248727.2 | XP_006248789.1 | 2 | 216 | |||
GenBank mRNA | AJ515244.1 | - | 2 | 170 | ||
BC087593.1 | - | 2 | 160 | 7310168 | ||
FM082155.1 | - | 2 | 172 | |||
FQ216277.1 | - | 2 | 163 |
Gene Symbol: LOC100125386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC081964.1 | - | 303 | 7125106 |