Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248575.2 | XP_006248637.1 | 40, 41 | 3675 | ||
XM_006248577.2 | XP_006248639.1 | 40, 41 | 3667 | |||
XM_006248578.2 | XP_006248640.1 | 40, 41 | 3699 | |||
XM_006248579.2 | XP_006248641.1 | 39, 40 | 3669 | |||
XM_008768830.1 | XP_008767052.1 | 40, 41 | 3874 | |||
XM_008768831.1 | XP_008767053.1 | 40, 41 | 3675 |