Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex11g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105902.1 | NP_001099372.1 | 2 | 263 | ||
XM_008768945.1 | XP_008767167.1 | 2 | 439 |