Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105920.1 | NP_001099390.1 | 1 | 94 | ||
GenBank mRNA | BC166568.1 | - | 1 | 77 | 7318881 |