Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006957.2 | NP_001006958.1 | 2 | 176 | ||
XM_006249134.1 | XP_006249196.1 | 2 | 399 | |||
GenBank mRNA | BC083818.1 | - | 2 | 168 | 7110979 | |
CB718613.1 | - | 2 | 176 |